Thiazole-2-carboxylic acid
Empirical Formula (Hill Notation):
C4H3NO2S
CAS Number:
Molecular Weight:
129.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing