Skip to Content
Merck
Thallium(III) trifluoroacetate technical grade

Thallium(III) trifluoroacetate

Thallium(III) trifluoroacetate technical grade
Synonym(s):
Thallic trifluoroacetate, Trifluoroacetic acid thallium(III) salt
Linear Formula:
(CF3COO)3Tl
CAS Number:
Molecular Weight:
543.43
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing