Skip to Content
Merck
Tetramethyltetraselenafulvalene 97%

Tetramethyltetraselenafulvalene

Tetramethyltetraselenafulvalene 97%
Synonym(s):
TMTSF
Empirical Formula (Hill Notation):
C10H12Se4
CAS Number:
Molecular Weight:
448.04

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing