![Terazosin for system suitability A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/214/89414c09-142c-496f-9009-99a4f6135976/640/89414c09-142c-496f-9009-99a4f6135976.png)
Terazosin for system suitability A
![Terazosin for system suitability A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/361/214/89414c09-142c-496f-9009-99a4f6135976/640/89414c09-142c-496f-9009-99a4f6135976.png)
Synonym(s):
Terazosin hydrochloride dihydrate, 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(tetrahydro-2-furanyl)carbonyl]piperazine hydrochloride dihydrate
Empirical Formula (Hill Notation):
C19H25N5O4 · HCl · 2H2O
CAS Number:
Molecular Weight:
459.92
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing