Skip to Content
Merck
(t-Bu)2PhP Pd G3 95%

(t-Bu)2PhP Pd G3

(t-Bu)2PhP Pd G3 95%
Synonym(s):
Methanesulfonato (di-tert-butyl) phenylphosphino (2′-amino-1,1′-biphenyl-2-yl) palladium(II), PPh(t-Bu)2-Pd-G3
Empirical Formula (Hill Notation):
C27H36NO3PPdS
CAS Number:
Molecular Weight:
592.04

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing