Sulfapyridine
Synonym(s):
4-Amino-N-[2-pyridyl]benzene sulfonamide, N1-(Pyridin-2-yl)sulfanilamide
Empirical Formula (Hill Notation):
C11H11N3O2S
CAS Number:
Molecular Weight:
249.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4