Skip to Content
Merck
SQ 22,536 ≥97% (HPLC), powder

SQ 22536

SQ 22,536 ≥97% (HPLC), powder
Synonym(s):
9-(Tetrahydro-2-furanyl)-9H-purin-6-amine, 9-THF-Ade
Empirical Formula (Hill Notation):
C9H11N5O
CAS Number:
Molecular Weight:
205.22

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing