Spironolactone
Synonym(s):
4-Pregnen-21-oic acid-17α-ol-3-one-7α-thiol γ-lactone 7-acetate, 7α-(Acetylthio)-17α-hydroxy-3-oxopregn-4-ene-21-carboxylic acid γ-lactone, Spironolactone
Empirical Formula (Hill Notation):
C24H32O4S
CAS Number:
Molecular Weight:
416.57
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing