![Spiraeoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/356/926/952a8880-7ffb-4055-82f5-e0c3895cf1c2/640/952a8880-7ffb-4055-82f5-e0c3895cf1c2.png)
Spiraeoside
![Spiraeoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/356/926/952a8880-7ffb-4055-82f5-e0c3895cf1c2/640/952a8880-7ffb-4055-82f5-e0c3895cf1c2.png)
Synonym(s):
3,3′,4′,5,7-Pentahydroxyflavone 4′-glucoside, Quercetin 4′-O-β-D-glucopyranoside, Quercetin 4′-glucoside, Spiraein, Spireoside
Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing