Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Spermine tetrahydrochloride
Synonym(s):
N,N′-Bis(3-aminopropyl)-1,4-butanediamine tetrahydrochloride
Empirical Formula (Hill Notation):
C
10
H
26
N
4
· 4HCl
CAS Number:
306-67-2
Molecular Weight:
348.18
EC Number:
206-189-8
Products
Product Category
bioactive small molecules (2)
Brand
Sigma-Aldrich (4)
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
85605
Spermine tetrahydrochloride,
BioUltra, for molecular biology, ≥99.5% (AT)
Expand
View Pricing
S1141
Spermine tetrahydrochloride,
BioReagent, for molecular biology
Expand
View Pricing
S2876
Spermine tetrahydrochloride,
powder or crystals, NMDA agonist/antagonist
Expand
View Pricing
85610
Spermine tetrahydrochloride,
≥99.0% (AT), powder or crystals
Expand
View Pricing