
Spermidine trihydrochloride

Synonym(s):
N-(3-Aminopropyl)-1,4-butanediamine trihydrochloride
Linear Formula:
NH2(CH2)3NH(CH2)4NH2 · 3HCl
CAS Number:
Molecular Weight:
254.63
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reference Material Type
Showing 1-4 of 4
Compare
Product Number
Description
Pricing