![Sparfloxacin ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/292/940/e05ed2bb-b862-42f5-8fec-495446fc19b5/640/e05ed2bb-b862-42f5-8fec-495446fc19b5.png)
Sparfloxacin
![Sparfloxacin ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/292/940/e05ed2bb-b862-42f5-8fec-495446fc19b5/640/e05ed2bb-b862-42f5-8fec-495446fc19b5.png)
Synonym(s):
5-Amino-1-cyclohexyl-7-(cis-3,5-dimethylpiperazino)-6,8-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid
Empirical Formula (Hill Notation):
C19H22F2N4O3
CAS Number:
Molecular Weight:
392.40
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing