Skip to Content
Merck
Sodium tert-pentoxide solution 1.4 M in THF

Sodium tert-pentoxide solution

Sodium tert-pentoxide solution 1.4 M in THF
Synonym(s):
tert-Amyl alcohol sodium salt, Sodium tert-amoxide
Empirical Formula (Hill Notation):
C5H11NaO
CAS Number:
Molecular Weight:
110.13

Products

Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing