![Sodium phosphite dibasic pentahydrate ≥98%](/deepweb/assets/sigmaaldrich/product/structures/109/127/3ec5bc99-ff3c-4d24-a16c-8220cd1a584f/640/3ec5bc99-ff3c-4d24-a16c-8220cd1a584f.png)
Sodium phosphite dibasic pentahydrate
![Sodium phosphite dibasic pentahydrate ≥98%](/deepweb/assets/sigmaaldrich/product/structures/109/127/3ec5bc99-ff3c-4d24-a16c-8220cd1a584f/640/3ec5bc99-ff3c-4d24-a16c-8220cd1a584f.png)
Synonym(s):
di-Sodium hydrogen phosphite
Linear Formula:
Na2HPO3 · 5H2O
CAS Number:
Molecular Weight:
216.04
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing