![Sodium Phosphate, Monobasic, Monohydrate, Molecular Biology Grade Useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers.](/deepweb/assets/sigmaaldrich/product/structures/465/012/7e3e8f4b-435f-47fc-a9eb-678713b66c56/640/7e3e8f4b-435f-47fc-a9eb-678713b66c56.png)
Sodium Phosphate, Monobasic, Monohydrate, Molecular Biology Grade
![Sodium Phosphate, Monobasic, Monohydrate, Molecular Biology Grade Useful in conjunction with Sodium Phosphate, Dibasic in the preparation of biological buffers.](/deepweb/assets/sigmaaldrich/product/structures/465/012/7e3e8f4b-435f-47fc-a9eb-678713b66c56/640/7e3e8f4b-435f-47fc-a9eb-678713b66c56.png)
Synonym(s):
Sodium phosphate monobasic monohydrate, Monosodium phosphate, Sodium dihydrogen phosphate monohydrate
Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
Products
Brand
Color
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1