
Sodium 2,3-dimercaptopropanesulfonate monohydrate

Synonym(s):
2,3-Dimercaptopropanesulfonic acid sodium salt monohydrate, DMPS
Linear Formula:
HSCH2CH(SH)CH2SO3Na · H2O
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1