Skip to Content
Merck
Sitagliptin Phosphate Pharmaceutical Secondary Standard; Certified Reference Material

Sitagliptin Phosphate

Sitagliptin Phosphate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Sitagliptin phosphate monohydrate, 7-[(3R)-3-amino-1-oxo-4-(2,4,5-trifluorophenyl)butyl]-5,6,7,8-tetrahydro-3-(trifluoromethyl)-1,2,4-triazolo[4,3-a]pyrazine phosphate, MK-0431
Empirical Formula (Hill Notation):
C16H15F6N5O · H3PO4 · H2O
CAS Number:
Molecular Weight:
523.32

Products

Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1