![SILu™MAB K1 Stable-Isotope Labeled Universal Monoclonal Antibody recombinant, expressed in CHO cells](/deepweb/assets/sigmaaldrich/product/images/224/033/b6c0e60c-df02-4ece-813e-4e47c40d512e/640/b6c0e60c-df02-4ece-813e-4e47c40d512e.jpg)
SILu™MAB K1 Stable-Isotope Labeled Universal Monoclonal Antibody
![SILu™MAB K1 Stable-Isotope Labeled Universal Monoclonal Antibody recombinant, expressed in CHO cells](/deepweb/assets/sigmaaldrich/product/images/224/033/b6c0e60c-df02-4ece-813e-4e47c40d512e/640/b6c0e60c-df02-4ece-813e-4e47c40d512e.jpg)
Synonym(s):
SILu™MAB Stable-Isotope Labeled Universal Monoclonal Antibody Standard human
Products
Product Category
Brand
Purity
Recombinant Host
Quality Segment
Package Size
Showing 1-1 of 1
Compare
Product Number
Description
Pricing