Skip to Content
Merck
(S)-(+)-Hexahydromandelic acid 98%

(S)-(+)-Hexahydromandelic acid

(S)-(+)-Hexahydromandelic acid 98%
Synonym(s):
(S)-α-Hydroxycyclohexaneacetic acid, (S)-(+)-Cyclohexylhydroxyacetic acid
Linear Formula:
C6H11CH(OH)CO2H
CAS Number:
Molecular Weight:
158.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing