Sabinyl acetate
Synonym(s):
cis-Sabinyl acetate, (1S,3R,5S)-1-Isopropyl-4-methylenebicyclo[3.1.0]hex-3-yl acetate, (1S,3R,5S)-4-Methylene-1-(1-methylethyl)bicyclo[3.1.0]hexan-3-ol 3-acetate, Sabinyl acetate
Empirical Formula (Hill Notation):
C12H18O2
CAS Number:
Molecular Weight:
194.27
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing