![(S)-(−)-5-Hydroxymethyl-2(5H)-furanone 98%](/deepweb/assets/sigmaaldrich/product/structures/170/412/cb8a3ac1-50ed-4044-91f1-2aa37cf9c021/640/cb8a3ac1-50ed-4044-91f1-2aa37cf9c021.png)
(S)-(−)-5-Hydroxymethyl-2(5H)-furanone
![(S)-(−)-5-Hydroxymethyl-2(5H)-furanone 98%](/deepweb/assets/sigmaaldrich/product/structures/170/412/cb8a3ac1-50ed-4044-91f1-2aa37cf9c021/640/cb8a3ac1-50ed-4044-91f1-2aa37cf9c021.png)
Empirical Formula (Hill Notation):
C5H6O3
CAS Number:
Molecular Weight:
114.10
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing