Skip to Content
Merck
Rutin trihydrate primary reference standard

Rutin trihydrate

Rutin trihydrate primary reference standard
Synonym(s):
Quercetin-3-rutinoside trihydrate, Vitamin P trihydrate
Empirical Formula (Hill Notation):
C27H30O16 · 3H2O
CAS Number:
Molecular Weight:
664.56
EC Number:

Products

Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3