Rivaroxaban for system suitability
Synonym(s):
(S)-Rivaroxaban, 5-Chloro-N-[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-2-thiophenecarboxamide
Empirical Formula (Hill Notation):
C19H18ClN3O5S
CAS Number:
Molecular Weight:
435.88
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing