Skip to Content
Merck
Rivaroxaban ≥98% (HPLC)

Rivaroxaban

Rivaroxaban ≥98% (HPLC)
Synonym(s):
(S)-Rivaroxaban, 5-Chloro-N-[[(5S)-2-oxo-3-[4-(3-oxo-4-morpholinyl)phenyl]-5-oxazolidinyl]methyl]-2-thiophenecarboxamide
Empirical Formula (Hill Notation):
C19H18ClN3O5S
CAS Number:
Molecular Weight:
435.88

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Research Area
Quality Segment
Application
API Family
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing