Skip to Content
Merck
Rink acid resin 100-200 mesh, extent of labeling: 0.5-1.5 mmol/g loading, 1 % cross-linked with divinylbenzene

Rink acid resin

Rink acid resin 100-200 mesh, extent of labeling: 0.5-1.5 mmol/g loading, 1 % cross-linked with divinylbenzene
Synonym(s):
(4-[(2,4-Dimethoxyphenyl)hydroxymethyl]phenol, polymer-bound, Rink acid-labile (hydroxymethyl)polystyrene

Products

Product Category
Brand
Quality Segment
Functional Group
Application
Cleavage Condition
Linker
Chain Type
Reaction Type