Skip to Content
Merck
Rhodamine 19 perchlorate Dye content ~95 %

Rhodamine 19 perchlorate

Rhodamine 19 perchlorate Dye content ~95 %
Synonym(s):
2-(6-Ethylamino-3-ethylimino-2,7-dimethyl-3H-xanthen-9-yl)-benzoic acid perchlorate, Rhodamine 575
Empirical Formula (Hill Notation):
C26H26N2O3 · HClO4
CAS Number:
Molecular Weight:
514.95
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing