Retrorsine
Synonym(s):
β-Longilobine, 12,18-Dihydroxysenecionan-11,16-dione
Empirical Formula (Hill Notation):
C18H25NO6
CAS Number:
Molecular Weight:
351.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing