![Retinyl acetate solution 100 μg/mL in ethanol (with 0.1% (w/v) BHT; degassed), reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/296/928/87b088d0-0f3b-4ac7-bcd4-d35a9fe421ac/640/87b088d0-0f3b-4ac7-bcd4-d35a9fe421ac.png)
Retinyl acetate solution
![Retinyl acetate solution 100 μg/mL in ethanol (with 0.1% (w/v) BHT; degassed), reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/296/928/87b088d0-0f3b-4ac7-bcd4-d35a9fe421ac/640/87b088d0-0f3b-4ac7-bcd4-d35a9fe421ac.png)
Synonym(s):
Vitamin A acetate solution
Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing