![Rebaudioside N phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/152/147/cde6ebef-3296-4347-bb06-f74ab4238171/640/cde6ebef-3296-4347-bb06-f74ab4238171.png)
Rebaudioside N
![Rebaudioside N phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/152/147/cde6ebef-3296-4347-bb06-f74ab4238171/640/cde6ebef-3296-4347-bb06-f74ab4238171.png)
Synonym(s):
Kaur-16-en-18-oic acid, 13-[(O-β-D-glucopyranosyl-(1→2)-O-[b-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]-, O-6-deoxy-α-L-mannopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl ester, (4α)-
Empirical Formula (Hill Notation):
C56H90O32
CAS Number:
Molecular Weight:
1275.29
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing