Rebaudioside A
Synonym(s):
(4α)-13-[(2-O-β-D-glucopyranosyl-3-O-β-Dglucopyranosyl-β-D-glucopyranosyl)-oxy]kaur-6-en-8-oic acid β-Dglucopyranosyl ester, Glycoside A3, Glycoside X, Stevioside a3
Empirical Formula (Hill Notation):
C44H70O23
CAS Number:
Molecular Weight:
967.01
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3