Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rat MCP-1 / CCL2 ELISA Kit
Products
Product Category
kits (2)
Brand
Sigma-Aldrich (2)
Gene Alias
Detection Method
Package Size
Species Reactivity
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
RAB0057
Rat MCP-1 / CCL2 ELISA Kit,
for serum, plasma and cell culture supernatant
Expand
View Pricing
RAB0058
Rat MCP-1 / CCL2 ELISA Kit,
for cell and tissue lysates
Expand
View Pricing