
Raloxifene Related Compound C

Synonym(s):
1-(2-{4-[6-Hydroxy-2-(4-hydroxyphenyl)benzothiophene-3-carbonyl]phenoxy}ethyl)piperidine 1-oxide
Empirical Formula (Hill Notation):
C28H27NO5S
CAS Number:
Molecular Weight:
489.58
Products
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing