Skip to Content
BR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quinine hydrochloride dihydrate
Synonym(s):
(8α, 9
R
)-6′-Methoxycinchonan-9-ol monohydrochloride dihydrate
Empirical Formula (Hill Notation):
C
20
H
24
N
2
O
2
· HCl · 2H
2
O
CAS Number:
6119-47-7
Molecular Weight:
396.91
EC Number:
205-001-1
Products
Product Category
alkaloids (1)
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Research Area
Quality Segment
Application
Mode of Action
Special Grade
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Q1125
Quinine hydrochloride dihydrate,
99-101% (titration), potassium channel blocker
Expand
View Pricing
22630
Quinine hydrochloride dihydrate,
BioReagent, suitable for fluorescence, ≥98.0% (NT)
Expand
View Pricing
22632
Quinine hydrochloride dihydrate,
tested according to Ph. Eur.
Expand
View Pricing