Skip to Content
Merck
PTAD-Alkyne 97%

PTAD-Alkyne

PTAD-Alkyne 97%
Synonym(s):
4-(4-(Prop-2-yn-1-yloxy)phenyl)-1,2,4-triazolidine-3,5-dione
Empirical Formula (Hill Notation):
C11H9N3O3
CAS Number:
Molecular Weight:
231.21

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing