Skip to Content
Merck
Prostaglandin B2 ≥98%, synthetic

Prostaglandin B2

Prostaglandin B2 ≥98%, synthetic
Synonym(s):
(5Z,13E,15S)-15-Hydroxy-9-oxoprosta-5,8(12),13-trien-1-oic acid, PGB2
Empirical Formula (Hill Notation):
C20H30O4
CAS Number:
Molecular Weight:
334.45

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing