Propionaldehyde-2,4-dinitrophenylhydrazone
Linear Formula:
C2H5CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
238.20
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing