Skip to Content
Merck
Propargylamine hydrochloride 95%

Propargylamine hydrochloride

Propargylamine hydrochloride 95%
Synonym(s):
2-Propynylamine hydrochloride, 3-Amino-1-propyne hydrochloride
Linear Formula:
HC≡CCH2NH2 · HCl
CAS Number:
Molecular Weight:
91.54
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing