Progesterone Related Compound H
Synonym(s):
Pregna-4,6-diene-3,20-dione
Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing