Skip to Content
Merck
Progesterone British Pharmacopoeia (BP) Reference Standard

Progesterone

Progesterone British Pharmacopoeia (BP) Reference Standard
Synonym(s):
4-Pregnene-3,20-dione
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Sterilization Method
Technique