
Procyclidine hydrochloride

Synonym(s):
α-Cyclohexyl-α-phenyl-1-pyrrolidinepropanol
Empirical Formula (Hill Notation):
C19H29NO · HCl
CAS Number:
Molecular Weight:
323.90
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Research Area
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing