Procainamide hydrochloride
Synonym(s):
Procainamide hydrochloride, 4-Amino-N-(2-diethylaminoethyl)benzamide hydrochloride, 4-Aminobenzoic acid 2-diethylaminoethylamide
Linear Formula:
H2NC6H4CONHCH2CH2N(C2H5)2·HCl
CAS Number:
Molecular Weight:
271.79
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3