Skip to Content
Merck
Prasugrel ≥98% (HPLC)

Prasugrel

Prasugrel ≥98% (HPLC)
Synonym(s):
5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate, CS 747, CS-747, Effient, Efient, LY640315
Empirical Formula (Hill Notation):
C20H20FNO3S
CAS Number:
Molecular Weight:
373.44

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing