Potassium Sorbate
Synonym(s):
Sorbic acid potassium salt, 2,4-Hexadienoic acid potassium salt, Potassium 2,4-hexadienoate, Potassium sorbate
Linear Formula:
CH3CH=CHCH=CHCOOK
CAS Number:
Molecular Weight:
150.22
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing