![Potassium phosphate tribasic monohydrate 90%](/deepweb/assets/sigmaaldrich/product/structures/101/830/c720b8d6-e944-4cc9-b7a9-012bdf7d38db/640/c720b8d6-e944-4cc9-b7a9-012bdf7d38db.png)
Potassium phosphate tribasic monohydrate
![Potassium phosphate tribasic monohydrate 90%](/deepweb/assets/sigmaaldrich/product/structures/101/830/c720b8d6-e944-4cc9-b7a9-012bdf7d38db/640/c720b8d6-e944-4cc9-b7a9-012bdf7d38db.png)
Synonym(s):
tert-Potassium phosphate monohydrate, Tripotassium phosphate
Empirical Formula (Hill Notation):
K3O4P · H2O
Products
Product Category
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing