Skip to Content
Merck
Poly(vinylpolypyrrolidone) ~110 μm particle size

Polyvinylpolypyrrolidone

Poly(vinylpolypyrrolidone) ~110 μm particle size
Synonym(s):
Polyvinylpolypyrrolidone, PVPP, Polyvinylpyrrolidone, cross-linked
CAS Number:

Products

Brand
Physical Form
Detection Method
Quality Segment
Application
Particle Size (µm)
Showing 1-2 of 2
Compare
Product Number
Description
Pricing