Polysorbate 80
Synonym(s):
TWEEN® 80, POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80
CAS Number:
Products
Product Category
Brand
Biological Source
Color
Feature
Physical Form
Product Line
Agency
Quality Segment
Application
Organoleptic Class
Package Size
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing