![Poly(ethylene glycol) methyl ether-block-poly(D,L lactide) PEG average Mn 5000, PDLA average Mn 50000](/deepweb/assets/sigmaaldrich/product/structures/125/868/d4649f54-57eb-44f4-a54d-0e46891a4123/640/d4649f54-57eb-44f4-a54d-0e46891a4123.png)
Poly(ethylene glycol) methyl ether-block-poly(D,L lactide)
![Poly(ethylene glycol) methyl ether-block-poly(D,L lactide) PEG average Mn 5000, PDLA average Mn 50000](/deepweb/assets/sigmaaldrich/product/structures/125/868/d4649f54-57eb-44f4-a54d-0e46891a4123/640/d4649f54-57eb-44f4-a54d-0e46891a4123.png)
Linear Formula:
HO[CH(CH3)COO]m[CH2CH2O]nCH3
Products
Brand
Color
Physical Form
Quality Segment
Optical Isomer
Polymer Composition
Polymer Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing