Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)
Synonym(s):
PEG-PPG-PEG
CAS Number:
Products
Brand
pH Value
Physical Form
Chemical Composition
Polymer Composition
Polymer Type
Showing 1-6 of 6
Compare
Product Number
Description
Pricing