Phosphorus pentachloride
Synonym(s):
Phosphorus pentachloride
Empirical Formula (Hill Notation):
Cl5P
CAS Number:
Molecular Weight:
208.24
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing