Skip to Content
Merck
Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit affinity isolated antibody, buffered aqueous solution

Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit

Peroxidase Anti-Peroxidase Soluble Complex antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Synonym(s):
PAP

Products

Product Category
Brand
Biological Source
Physical Form
Antibody Form
Post Translational Modification
Technique